Erlenmeyer flasks with standard ground joints, DURAN®
250 ml, 140 mm, 85 mm, 29/32 NS
Catalog # SCOT241933701
Supplier: DWK Life Sciences
Erlenmeyer flasks with standard ground joints, DURAN®
Catalog # SCOT241933701
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduations and labelling area.
According to DIN ISO 4797
*non-DIN ISO size
Specifications
- Ext.Ø:85 mm
- Socket size:29/32 NS
- Capacity:250 ml
- Height:140 mm



