Order Entry
Italy
ContactUsLinkComponent
I 800 BNC,COMBINED IODIDE SELECTIVE ELECTRODE, WITH 3.3 FT (1 M) CABLE, BNC PLUG, 0.006 TO 127,000 MG/L, REQUIRES ELY/BR/503 REFEREN CE ELECTROLYTE, IONIC STRENGTH ADAPTER ISA/FK, STANDARD SOLUTION ES/I DELIVERY CONTAINS ELECTROLYTE 1 * 1 items
  662-7380
 :  WTW
I 800 BNC,COMBINED IODIDE SELECTIVE ELECTRODE, WITH 3.3 FT (1 M) CABLE, BNC PLUG, 0.006 TO 127,000 MG/L, REQUIRES ELY/BR/503 REFEREN CE ELECTROLYTE, IONIC STRENGTH ADAPTER ISA/FK, STANDARD SOLUTION ES/I DELIVERY CONTAINS ELECTROLYTE 1 * 1 items
  662-7380
 :  WTW
Frequently Bought Together