Order Entry
Italy
ContactUsLinkComponent
PB 500 DIN,LEAD ION SELECTIVE ELECTRODE WITH 3.3 FT (1 M) CABLE, DIN PLUG, 0.2 TO 20,000 MG/L, REQUIRES REFERENCE R 503/D OR /P WITH ELY/BR/503, IONIC STRENGTH ADAPTER ISA/FK, STANDARD SOLUTION ES/PB. ACCESSORIES ARE NOT INCLUDED IN DELIVERY 1 * 1 items
Codice catalogo 662-0269
Supplier:  WTW
PB 500 DIN,LEAD ION SELECTIVE ELECTRODE WITH 3.3 FT (1 M) CABLE, DIN PLUG, 0.2 TO 20,000 MG/L, REQUIRES REFERENCE R 503/D OR /P WITH ELY/BR/503, IONIC STRENGTH ADAPTER ISA/FK, STANDARD SOLUTION ES/PB. ACCESSORIES ARE NOT INCLUDED IN DELIVERY 1 * 1 items
Codice catalogo 662-0269
Supplier:  WTW
Frequently Bought Together