Order Entry
Italy
Orders LinkContactUsLinkComponent
Joint clips for conical joints, POM, Quickfit®
Joint clips for conical joints, POM, Quickfit®
Blue, 19/26
Catalog # 201-8567
Supplier:   SciLabware
Joint clips for conical joints, POM, Quickfit®
Catalog # 201-8567
Supplier:  SciLabware
Joint clips for conical joints, POM, Quickfit®
Joint clips for conical joints, POM, Quickfit®
Blue, 19/26
Price
53,20 €
Log in for availability
1 * 10 pezzi
Product Details & Documents
POM, coloured. Polyacetal clips for retaining glassware having Quickfit® conical joints.
  • Easy to fit and remove
  • Can be used at temperatures up to +80 °C
  • Clips are colour coded for easy identification
Specifications
  • For NS:
    19/26
  • Colour:
    Blue
  • Seq:
    04
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..