Order Entry
Italy
Orders LinkContactUsLinkComponent
Joint clips for conical joints, POM, Quickfit®
Purple, 12/21
Catalog # 201-8186
Supplier:   SciLabware
Joint clips for conical joints, POM, Quickfit®
Catalog # 201-8186
Supplier:  SciLabware
Joint clips for conical joints, POM, Quickfit®
Joint clips for conical joints, POM, Quickfit®
Purple, 12/21
Price
50,10 €
Log in for availability
1 * 10 pezzi
Product Details & Documents
POM, coloured. Polyacetal clips for retaining glassware having Quickfit® conical joints.
  • Easy to fit and remove
  • Can be used at temperatures up to +80 °C
  • Clips are colour coded for easy identification
Specifications
  • For NS:
    12/21
  • Colour:
    Purple
  • Seq:
    02