VWR®, Speciality pH Electrodes
Flat extra wide, double junction (HDPE/Pellon), +10...+100 °C, Epoxy, Gel, 15×115 mm, BNC, Agar, microbiological samples, cheese, butter, water
Supplier: VWR Collection
Jump to:
Product Details & Documents
With 1 m fixed cable.
pH range: 0 to 14
Specifications
- Connection:BNC
- Ø×L:15×115 mm
- For:Agar, microbiological samples, cheese, butter, water
- Type:Flat extra wide, double junction (HDPE/Pellon)
- Material:Epoxy
- Seq:02
- Electrolyte:Gel
- Temp.:+10...+100 °C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



