Safety spectacles, Op-Tema®
Anti-fog, Black, Clear PC
111-0731
Comfortable safety spectacles with lightweight full view lens and built-in moulded browbar.
- Soft, adjustable nose piece
- Fully adjustable
- Adjustable, break-away Flexicord retainer for added safety
Weight: 28 g
Certifications: EN 166, EN 170; 1F
- Type:Anti-fog
- Colour:Black
- Seq:05
- Lenses:Clear PC