Coagulation tubes, BD Vacutainer®
Blood Collection tube, BD Vacutainer®, Plastic, BD Hemogard™, Sodium citrate (0,109 M), 75 mm, 13 mm, Light blue, 1,8 ml
Catalog # BDAM363047
Jump to:
Discontinued
Product Details & Documents
PET/PP or glass.
- With buffered citrate
- With paper lable
- Hemogard™ closure
Citrate tubes
Specifications
- Closure type:BD Hemogard™
- Ø:13 mm
- Volume:1,8 ml
- Description:Blood Collection tube, BD Vacutainer®
- Length:75 mm
- Material:Plastic
- Cap colour:Light blue
- Additive:Sodium citrate (0,109 M)



