Ion selective half-cell electrodes
1×10-6...1 mol/l, 2 - 13, 0...+80 °C, DX280-Br-, 1 mol/l KNO₃, Br-
Catalog # 662-2802
Supplier: Mettler - Toledo
Jump to:
Product Details & Documents
These ion selective half cells with solid-state membrane allow a quick and easy determination of concentration and activity of a variety of dissolved ions and gases.
- Modular and versatile ion selective half cells consisting of a universal shaft and a ion specific, exchangeable membrane
- Conform to RoHS (‘Restriction of Hazardous Substances’ in electrical and electronic equipment) and WEEE (‘Waste Electrical and Electronic Equipment’)
: Membrane kits available separately.
Specifications
- Concentration:1×10-6...1 mol/l
- For:Br-
- Type:DX280-Br-
- Seq:01
- Electrolyte:1 mol/l KNO₃
- Temp.:0...+80 °C
- pH:2 - 13
Accessories
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX280-Br⁻
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX240-Ca²⁺
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX239-K⁺
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX262-NO₃⁻
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX235-Cl⁻
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX219-F⁻
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX218-NH₄⁺



