Order Entry
Italy
Orders LinkContactUsLinkComponent
Ion selective half-cell electrodes

Ion selective half-cell electrodes

1×10-6...1 mol/l, 2 - 12, 0...+50 °C, DX239-K⁺, 2 mol/l MgSO₄, K⁺
Catalog # 662-2806
Supplier:   Mettler - Toledo
Ion selective half-cell electrodes
Catalog # 662-2806
Supplier:  Mettler - Toledo
Ion selective half-cell electrodes
Ion selective half-cell electrodes

1×10-6...1 mol/l, 2 - 12, 0...+50 °C, DX239-K⁺, 2 mol/l MgSO₄, K⁺

Price
1 220,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

These ion selective half cells with solid-state membrane allow a quick and easy determination of concentration and activity of a variety of dissolved ions and gases.

  • Modular and versatile ion selective half cells consisting of a universal shaft and a ion specific, exchangeable membrane
  • Conform to RoHS (‘Restriction of Hazardous Substances’ in electrical and electronic equipment) and WEEE (‘Waste Electrical and Electronic Equipment’)

Ordering information: Membrane kits available separately.

Documents

Specifications
  • Concentration:
    1×10-6...1 mol/l
  • For:
    K⁺
  • Type:
    DX239-K⁺
  • Seq:
    05
  • Electrolyte:
    2 mol/l MgSO₄
  • Temp.:
    0...+50 °C
  • pH:
    2 - 12
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Membrane kit for DX280-Br⁻
    Membrane kit for DX280-Br⁻
    662-2767
    Log in for availability
    1 * 1 pezzi - 675,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Membrane kit for DX240-Ca²⁺
    Membrane kit for DX240-Ca²⁺
    662-2770
    Log in for availability
    1 * 1 pezzi - 675,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Membrane kit for DX239-K⁺
    Membrane kit for DX239-K⁺
    662-2771
    Log in for availability
    1 * 1 pezzi - 665,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Membrane kit for DX262-NO₃⁻
    Membrane kit for DX262-NO₃⁻
    662-2772
    Log in for availability
    1 * 1 pezzi - 690,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Membrane kit for DX235-Cl⁻
    Membrane kit for DX235-Cl⁻
    662-2768
    Log in for availability
    1 * 1 pezzi - 675,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Membrane kit for DX219-F⁻
    Membrane kit for DX219-F⁻
    662-2769
    Log in for availability
    1 * 1 pezzi - 680,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Membrane kit for DX218-NH₄⁺
    Membrane kit for DX218-NH₄⁺
    662-2773
    Log in for availability
    1 * 1 pezzi - 675,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.