Cylinders, without lid
120 mm
Catalog # HECH44409030
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Jars, made of glass or stainless steel, with base, without lids.
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
: Lids with knob available must be ordered separately.
Specifications
- Ext.Ø:120 mm
- Seq:19