Order Entry
Italy
Orders LinkContactUsLinkComponent
Double spatulas, micro, stiff

Double spatulas, micro, stiff

210 mm, 5 mm
Catalog # RSGA835.210
Supplier:   RSG
Double spatulas, micro, stiff
Catalog # RSGA835.210
Supplier:  RSG
Double spatulas, micro, stiff
Double spatulas, micro, stiff

210 mm, 5 mm

Price
30,40 €
Log in for availability
1 * 5 pezzi
Product Details & Documents

Rigid micro double spatulas made of stainless steel.

Specifications
  • Length:
    210 mm
  • Blade width:
    5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.