Accessories for heating circulators
2 Barbed fittings, M10×1, for 8 mm inner diameter tube, ED, EH, MA, ME
Catalog # JULA8970460
Supplier: Julabo GmbH
Jump to:
Product Details & Documents
Specifications
- Description:2 Barbed fittings, M10×1, for 8 mm inner diameter tube
- Seq:07
- For:ED, EH, MA, ME