Accessories for heating circulators
2× adapters, M16×1 female to NPT ½" male
Catalog # JULA8890008
Supplier: Julabo GmbH
Jump to:
Product Details & Documents
Specifications
- Description:2× adapters, M16×1 female to NPT ½" male