Order Entry
Italy
ContactUsLinkComponent
Areometer, BS718, M50 series SP precision, measuring range:0.85-0.90:0.001g/ml, without thermometer, length:270mm, reference tempera ture:15°C, suitable for government verification. 1 * 1 items
Catalog # AMARH808110
Nome del fornitore:  Amarell
Areometer, BS718, M50 series SP precision, measuring range:0.85-0.90:0.001g/ml, without thermometer, length:270mm, reference tempera ture:15°C, suitable for government verification. 1 * 1 items
Catalog # AMARH808110
Nome del fornitore:  Amarell
Frequently Bought Together