Order Entry
Italy
Orders LinkContactUsLinkComponent
TLC introductory kit, Micro Set A
TLC introductory kit, Micro Set A
Collection of 4 indiviudual components of test dye mixture 1
Codice catalogo MANA814011
Nome del fornitore:   MACHEREY-NAGEL
TLC introductory kit, Micro Set A
Codice catalogo MANA814011
Nome del fornitore:  MACHEREY-NAGEL
TLC introductory kit, Micro Set A
TLC introductory kit, Micro Set A
Collection of 4 indiviudual components of test dye mixture 1
Price
48,60 €
Log in for availability
1 * 1 KIT
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
An ideal introductory kit for the science education sector. The kit contains all the the chemicals, plates and accessories for students to perform separations of dyes including: anthaquinone dye mixtures, lipophilic dye mixtures and food dye mixtures. The TLC plates are polyester sheet based with either silica, cellulose or alumina as coating.
Delivery information: Kit includes 3 developing tanks, 50 glass capillaries (1 µl), 1 spotting guide, 1×10 ml measuring cylinder, 50 sheets 40×80 mm POLYGRAM SIL G/UV₂₅₄, 50 sheets 40×80 mm ALOX N/UV₂₅₄ and 50 sheets 40×80 mm of CEL 300, 3 dye mixtures (8 ml), 3 prepared solvent mixtures (100 ml), 2 felt pens and instruction manual.
Specifications
  • Description:
    Collection of 4 indiviudual components of test dye mixture 1
  • Seq:
    02