Order Entry
Italy
Orders LinkContactUsLinkComponent
VISOCOLOR® Colorimetric Test, Copper

VISOCOLOR® Colorimetric Test, Copper

VISOCOLOR® HE Copper, refill pack, Reagents for 150 tests in a box, 0,0 • 0,04 • 0,07 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 • 0,40 • 0,50 mg/L (Cu²⁺), Cuprizon, Visual, 233×129×45 mm (Including packaging), 150, 1 KIT
Catalog # MANA920150
Supplier:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Copper
Catalog # MANA920150
Supplier:  MACHEREY-NAGEL
CAS Number:  
VISOCOLOR® Colorimetric Test, Copper
VISOCOLOR® Colorimetric Test, Copper

VISOCOLOR® HE Copper, refill pack, Reagents for 150 tests in a box, 0,0 • 0,04 • 0,07 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 • 0,40 • 0,50 mg/L (Cu²⁺), Cuprizon, Visual, 233×129×45 mm (Including packaging), 150, 1 KIT

CAS Number:  
Price
76,80 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Reagents for the determination of copper in water samples.

  • Combines simple implementation with high measurement reliability

Simple determination directly on site or in the laboratory.

Sea water analysis: Yes

Hazardous material: No

Specifications

Specifications

  • Chemical method:
    Cuprizon
  • Pk:
    1 KIT
  • Description:
    VISOCOLOR® HE Copper, refill pack
  • Dimensions:
    233×129×45 mm (Including packaging)
  • For unit:
    Visual
  • Tests per kit:
    150
  • Range:
    0,0 • 0,04 • 0,07 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 • 0,40 • 0,50 mg/L (Cu²⁺)
  • Contents:
    Reagents for 150 tests in a box
  • Cat. No.:
    MANA920150
  • Platform:
    Colorimetric test kits
  • Shelf life (months):
    24
  • Storage temperature:
    15...25 °C
  • Weight:
    276,02 g (Including packaging)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.