VISOCOLOR® Colorimetric Test, Copper
VISOCOLOR® HE Copper, Reagents for 150 tests, color chart and accessories in a box, 0,0 • 0,04 • 0,07 • 0,10 • 0,15 • 0,20 • 0.25 • 0,30 • 0,40 • 0,50 mg/L (Cu²⁺), Cuprizon, Visual, 245×145×47 mm (Including packaging), 150, 1 KIT
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Reagents for the determination of copper in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: No
Specifications
Specifications
- Chemical method:Cuprizon
- Pk:1 KIT
- Description:VISOCOLOR® HE Copper
- Dimensions:245×145×47 mm (Including packaging)
- For unit:Visual
- Tests per kit:150
- Range:0,0 • 0,04 • 0,07 • 0,10 • 0,15 • 0,20 • 0.25 • 0,30 • 0,40 • 0,50 mg/L (Cu²⁺)
- Contents:Reagents for 150 tests, color chart and accessories in a box
- Cat. No.:MANA920050
- Platform:Colorimetric test kits
- Shelf life (months):24
- Storage temperature:15...25 °C
- Weight:520 g (Including packaging)



