Order Entry
Italy
Orders LinkContactUsLinkComponent
VISOCOLOR® Colorimetric Test, Ammonia

VISOCOLOR® Colorimetric Test, Ammonia

VISOCOLOR® HE Ammonium, Reagents for 110 tests, colour chart and accessories in a box, 0,0 • 0,02 • 0,04 • 0,07 • 0,10 • 0,15 • 0,20 • 0,30 • 0,40 • 0,50 mg/L (NH₄⁺), Indophenol, Visual, 239×135×47 mm (Including packaging), Sea water non compatible, 110, 1 KIT
Catalog # MANA920006
Supplier:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Ammonia
Catalog # MANA920006
Supplier:  MACHEREY-NAGEL
CAS Number:  
VISOCOLOR® Colorimetric Test, Ammonia
VISOCOLOR® Colorimetric Test, Ammonia

VISOCOLOR® HE Ammonium, Reagents for 110 tests, colour chart and accessories in a box, 0,0 • 0,02 • 0,04 • 0,07 • 0,10 • 0,15 • 0,20 • 0,30 • 0,40 • 0,50 mg/L (NH₄⁺), Indophenol, Visual, 239×135×47 mm (Including packaging), Sea water non compatible, 110, 1 KIT

CAS Number:  
Price
124,00 €
Log in for availability
1 * 1 KIT
Specifications

Specifications

  • Chemical method:
    Indophenol
  • Pk:
    1 KIT
  • Description:
    VISOCOLOR® HE Ammonium
  • Dimensions:
    239×135×47 mm (Including packaging)
  • Type:
    Sea water non compatible
  • For unit:
    Visual
  • Tests per kit:
    110
  • Range:
    0,0 • 0,02 • 0,04 • 0,07 • 0,10 • 0,15 • 0,20 • 0,30 • 0,40 • 0,50 mg/L (NH₄⁺)
  • Contents:
    Reagents for 110 tests, colour chart and accessories in a box
  • Cat. No.:
    MANA920006
  • Platform:
    Colorimetric test kits
  • Model:
    MANA920006
  • Shelf life (months):
    12
  • Storage temperature:
    15...25 °C
  • Weight:
    331,92 g (Including packaging)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.