SPE columns, CHROMABOND® Easy
Easy, PP, 50 Å, 80 µm, 15 ml, 500 mg
Catalog # MANA730757
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
CHROMABOND® Easy is a polar, bifunctionally modified polystyrene-divinylbenzene copolymer for SPE.
- Applicable for a wide range of analytes without preconditioning
- More hydrophilic than conventional polystyrene-divinylbenzene polymers
- Easily wettable with water
Applications: Polar pesticides (acidic, neutral, basic) or phenols from water; drug analysis from urine, blood, serum or from tablets and creams.
Specifications
- Particle Size:80 µm
- Pore size:50 Å
- Capacity:15 ml
- Seq:10
- Bed weight:500 mg
- Column hardware:PP
- Phase:Easy
- Cat. No.:MANA730757
- pH range:1 - 14
- Surface area:650 - 700 m²/g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



