Order Entry
Italy
ContactUsLinkComponent
EC HPLC column EC 125/4 NUCLEOSIL 100-5 C8 HD Length: 125 mm, ID: 4 mm pack of 1 1 * 1 items
Catalog # MANA720195.40
Nome del fornitore:  MACHEREY-NAGEL
EC HPLC column EC 125/4 NUCLEOSIL 100-5 C8 HD Length: 125 mm, ID: 4 mm pack of 1 1 * 1 items
Catalog # MANA720195.40
Nome del fornitore:  MACHEREY-NAGEL
Frequently Bought Together