cirfi MN 605, 7,0 cm 1 * 100 pezzi
Catalog # MANA423007
Supplier: MACHEREY-NAGEL
Frequently Bought Together
Guanti in nitrile, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Pipette sierologiche, Standard Line
Supplier: VWR Collection
VWR®, Guanti in Nitrile
Supplier: VWR Collection
VWR®, Salviette per il viso
Supplier: VWR Collection
Strisce reattive pH, MQuant®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, pipette di trasferimento, alte prestazioni
Supplier: VWR Collection
VWR® NITRILE LIGHT, Guanti in nitrile
Supplier: VWR Collection
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
Supplier: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals