Order Entry
Italy
Orders LinkContactUsLinkComponent
Base materials

Base materials

Tryptone Biochemica, 1 kg
Catalog # APPCA1553.1000
Supplier:   PanReac AppliChem
Base materials
Catalog # APPCA1553.1000
Supplier:  PanReac AppliChem
CAS Number:  
Base materials
Base materials

Tryptone Biochemica, 1 kg

CAS Number:  
Price
168,00 €
Log in for availability
1 * 1 kg
Product Details & Documents

Components of many microbiological growth media.

  • Store in a dry place
Specifications

Specifications

  • Pk:
    1 kg
  • Description:
    Tryptone Biochemica

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.