Inoculating loops with needle end
Inoculating loops, loop at both ends, flexible, 1+10 µl, Yellow, 50 bags with 20 pieces
Supplier: Brand
Jump to:
Product Details & Documents
PS, gamma sterilised, disposable. For inoculation of nutrient media. Available either with loop at one end and needle at the other or loops at both ends. High flexibility permits gentle streaking without damaging the nutrient surface.
Documents
Specifications
- Packed:50 bags with 20 pieces
- Capacity:1+10 µl
- Colour:Yellow
- Description:Inoculating loops, loop at both ends, flexible