Order Entry
Italy
ContactUsLinkComponent
Analytical balance, with type approval - Weighing-range 1 in g : 220 - Read-out 1 in g : 0.0001 - Weighing- plate in mm : 80 1 * 1 items
Catalog # KERNABJ220-4M
Supplier:  KERN SOHN
Analytical balance, with type approval - Weighing-range 1 in g : 220 - Read-out 1 in g : 0.0001 - Weighing- plate in mm : 80 1 * 1 items
Catalog # KERNABJ220-4M
Supplier:  KERN SOHN
Frequently Bought Together