Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Dropping bottles, glass, Wheaton®

with ground glass stopper, 100 ml, 120 mm, 56 mm
  WHEAW211735
Dropping bottles, glass, Wheaton®
  WHEAW211735
Dropping bottles, glass, Wheaton®
Dropping bottles, glass, Wheaton®

with ground glass stopper, 100 ml, 120 mm, 56 mm

 
270,00 €
   
1 * 6 pezzi
 

Manufactured from soda-lime glass material.

  • Available in clear or amber glass format
  • Amber coloured to protect light sensitive substanzes
  • Not autoclavable

Dropping bottles delivers stains or indicator solutions drop by drop. Complete with a glass stopper, a partial turn of the ground glass stopper seals the bottle.

 : Material conforms to USP Type III requirements.

 
  • Ext.Ø:
    56 mm
  • Type:
    with ground glass stopper
  • Capacity:
    100 ml
  • Height:
    120 mm