Inserisci ordine
Italy
Orders LinkContactUsLinkComponent
BOD bottles, WHEATON®
BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 313-336, 300 ml
Catalog # WHEA227497-14G
Supplier:   DWK Life Sciences
BOD bottles, WHEATON®
Catalog # WHEA227497-14G
Supplier:  DWK Life Sciences
BOD bottles, WHEATON®
BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 313-336, 300 ml
Price
787,00 €
Log in for availability
1 * 24 pezzi
Product Details & Documents
Made of Type I borosilicate glass.
  • Available with or without stopper
  • With white labeling area
Specifications
  • Capacità:
    300 ml
  • Descrizione:
    BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 313-336