IGM (µ HEAVY CHAIN SPECIFIC) 0.5 MG 1 * 0,5 mg
Catalog # SOUT2022-01
Supplier: SOUTHERN BIOTECHNOLOGY
IGM (µ HEAVY CHAIN SPECIFIC) 0.5 MG 1 * 0,5 mg
Catalog # SOUT2022-01
Supplier: SOUTHERN BIOTECHNOLOGY
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Dimethyl sulphoxide ≥99.5%, BioChemica® cell culture reagent
Supplier: PanReac AppliChem
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
G418 disulphate in solution
Supplier: PanReac AppliChem
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF