Dressing forceps, micro
Curved, fine, 105 mm
HAMMHWC201-10
WIRONIT steel (18/12 stainless steel with 18% chrome and 10 to 12% nickel).
- Resistant to corrosion
- Anti-magnetic
- Serrated
- Type:Curved, fine
- Length:105 mm
- Seq:06
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



