Order Entry
Italy
Orders LinkContactUsLinkComponent
Evaporating dishes, flat shape

Evaporating dishes, flat shape

50 ml, 20 mm, 60 mm
Catalog # BOCH8570
Supplier:   BOCHEM
Evaporating dishes, flat shape
Catalog # BOCH8570
Supplier:  BOCHEM
Evaporating dishes, flat shape
Evaporating dishes, flat shape

50 ml, 20 mm, 60 mm

Price
24,50 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Evaporating dishes are made of stainless steel 18/10.

  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean
Specifications
  • Ext.Ø:
    60 mm
  • Capacity:
    50 ml
  • Height:
    20 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.