Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Plastic drum, 10 L
Supplier: VWR Chemicals
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Catalog # 9713.9010
Supplier: VWR Chemicals
CAS Number: 50-00-0
Jump to:
Product Details & Documents
(= 10% Formaline solution)
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- :Stabilised with methanol 0,5-1,5 %
- Storage Temperature:Ambient
- EINECS:200-001-8
Certificates
Search by Lot Number10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 25L Plastic drum for liquids DIN 61 (ZPACK13952500)
Specifications
Specification Test Results
- Assay (Formaldehyde):3.5 - 4.5 %
- pH (20 °C):6.9 - 7.1
- Methanol:0.5 - 1.5 %
Specifications
- Pack type:Plastic drum
- Pk:10 L



