Order Entry
Italy
Orders LinkContactUsLinkComponent
Soda lime, with indicator

Soda lime, with indicator

Plastic bottle, 500 g
Catalog # 1.06733.0501
Corrosive
Irritant
Supplier:   Merck
Soda lime, with indicator
Catalog # 1.06733.0501
Supplier:  Merck
CAS Number:  8006-28-8
Soda lime, with indicator

Plastic bottle, 500 g

CAS Number:  8006-28-8
Price
127,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:8006-28-8
  • MDL Number:MFCD00134094
  • Storage Temperature:Ambient
  • ADR:8,III
  •  :M-Clarity™ quality level = MQ200
  • UN:1907
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.