COLUMN HXL-L GUARDCOLUMN, 7 UM, FOR G1000HXL-G4000HXL AND GMHXL-L - POLYMER - 6MM ID X 4CM L 1 * 1 items
Catalog # TOYO0007113
Supplier: TOSOH Bioscience
COLUMN HXL-L GUARDCOLUMN, 7 UM, FOR G1000HXL-G4000HXL AND GMHXL-L - POLYMER - 6MM ID X 4CM L 1 * 1 items
Catalog # TOYO0007113
Supplier: TOSOH Bioscience
Frequently Bought Together
Glass screw top headspace vials, 10 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
Safety canisters
Supplier: S.C.A.T.
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, TLC Glass Plates, Silica, Unmodified Layers
Supplier: VWR Collection
Screw neck vials, N 15
Supplier: MACHEREY-NAGEL
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
VWR® ACE® Excel® C18 HPLC/UHPLC Columns, Analytical, 3 µm
Supplier: Avantor
Screw caps, 9 mm, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Collecting trays, b.safe
Supplier: Bohlender
Exhaust filters, V3.0
Supplier: S.C.A.T.
Digestion tubes, DigiTUBE®
Supplier: PerkinElmer
Connection Pipe Set
Supplier: TOSOH Bioscience
Safety caps and safety waste cap accessories and replacement parts
Supplier: S.C.A.T.
COLUMN G2000HXL, 5 UM, 20A - POLYMER - 7.8MM ID X 30CM L 1 * 1 items
Supplier: TOSOH Bioscience
SPME fiber assemblies, Supelco®
Supplier: Merck