Jump to:
Product Details & Documents
Synonyms: p-Chlorophenol
- CAS Number:106-48-9
- Formula:C₆H₅ClO
- MDL Number:MFCD00002318
- Molecular Weight:128.56 g/mol
- Merck Index:13,02173
- Boiling Pt:220 °C (1013 hPa)
- Melting Pt:41…44 °C
- Density:1.31 g/cm³ (20 °C)
- Flash Pt:121 °C
- Storage Temperature:Ambient
- UN:2020
- ADR:6.1,III
- :M-Clarity™ quality level = MQ200
- EINECS:203-402-6
Documents
Specifications
Specification Test Results
- Solubility:27 g/l (20 °C)
- Melting Point:41 - 44 °C
- Molar Mass:128.55 g/mol
- Boiling Point:220 °C (1013 hPa)
- Vapor Pressure:0.15 hPa (20 °C)
- Flash Point:121 °C
- Density:1.26 g/cm3 (45 °C)
- Synonyms:4-Chloro-1-hydroxybenzene
- Hill Formula:C6H5ClO
- Chemical Formula:4-(Cl)C6H4OH
Specifications
- Pack type:Plastic bottle for solids
- Pk:5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



