Flasks, round bottom, Quickfit® ground joints
50 ml, 24/29 NS, 95 mm
Catalog # 201-8533
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
: ISO 1773, ISO 4797.
Specifications
- Socket size:24/29 NS
- Capacity:50 ml
- Height:95 mm



