Order Entry
Italy
Orders LinkContactUsLinkComponent
Forceps
Forceps
Fine tip, White, 145 mm
Catalog # LPIT291106
Supplier:   LP ITALIANA
Forceps
Catalog # LPIT291106
Supplier:  LP ITALIANA
Forceps
Forceps
Fine tip, White, 145 mm
Price
110,00 €
Log in for availability
1 * 10 pezzi
Product Details & Documents
White forceps made of PMP (Polymethylpentene).
Specifications
  • Type:
    Fine tip
  • Colour:
    White
  • Length:
    145 mm