Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Tissue processing/embedding cassettes, Histosette® II, M485

Orange
Catalog # 720-0224
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Histosette® II, M485
Catalog # 720-0224
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Histosette® II, M485
Tissue processing/embedding cassettes, Histosette® II, M485

Orange

Price
111,00 €
Log in for availability
1 * 1.000 pezzi
Product Details & Documents

Acetal polymer, similar to M492, but covers are already removed from cassettes and are packed separately in the case.

  • Convenient for cassette labelling instruments as the covers are already removed
  • Resistant to the chemical action of histological solvents
  • Flow-through slots maximise fluid exchange
Specifications
  • Colour:
    Orange