Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Quality control test suspensions

Clostridium sporogenes, 10 mL
Catalog # NAMSGP-02
Supplier:   CROSSTEX
Quality control test suspensions
Catalog # NAMSGP-02
Supplier:  CROSSTEX
CAS Number:  
Quality control test suspensions

Clostridium sporogenes, 10 mL

CAS Number:  
Price
376,00 €
Log in for availability
1 * 10 mL
Product Details & Documents

The control suspension (growth promotion) contains <100 CFU`s/0,1 ml.

  • 10 ml pro vial
  • 100 dose

 

Specifications

Specifications

  • Pk:
    10 mL
  • Description:
    Clostridium sporogenes

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.