Stoppers, press down, VacuCap®
Grey, 16 mm
Catalog # 720-0553
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Ideal closures for glass and plastic reagent, sample and culture tubes, LDPE.
- Disposable seals are an inexpensive alternative to standard seals
- Double flanged seal for an impermeable, secure fit
- Colour coded for sample identification
Flexible to protect from aerosols or highly infectious microorganisms. Samples are protected from cross-contamination and the work environment from infection due to leaks. The double flanged seal guarantees a tight and impermeable fit for a wide range of applications, including centrifuging.
Specifications
- Ø bottom:16 mm
- Colour:Grey