Order Entry
Italy
Orders LinkContactUsLinkComponent
Ammonia solution ∼32%, GPR RECTAPUR®

Ammonia solution ∼32%, GPR RECTAPUR®

Plastic bottle, fluorinated, 5 L
Catalog # 21192.367
Corrosive
Irritant
Environment
Supplier:   VWR Chemicals
Ammonia solution ∼32%, GPR RECTAPUR®
Catalog # 21192.367
Supplier:  VWR Chemicals
CAS Number:  1336-21-6
Ammonia solution ∼32%, GPR RECTAPUR®
Ammonia solution ∼32%, GPR RECTAPUR®

Plastic bottle, fluorinated, 5 L

CAS Number:  1336-21-6
Price
97,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.88 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specification Test Results

  • Assay:
    28 - 33 %
  • Density (20/4):
    0.880 - 0.910
  • Evaporation residue:
    Max. 100 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • CO3 (Carbonate):
    Max. 100 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • Fe (Iron):
    Max. 1 ppm
  • Pb (Lead):
    Max. 1 ppm

Specifications

  • Pack type:
    Plastic bottle, fluorinated
  • Pk:
    5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.