Ammonia solution ∼32%, GPR RECTAPUR®
Glass bottle, 1 L
Supplier: VWR Chemicals
Ammonia solution ∼32%, GPR RECTAPUR®
Catalog # 21192.298
Supplier: VWR Chemicals
CAS Number: 1336-21-6
Jump to:
Product Details & Documents
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- Boiling Pt:32…37.7 °C (1013 hPa)
- Melting Pt:–60…–57.5 °C
- Density:0.88 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-647-6
- UN:2672
- ADR:8,III
Documents
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415250) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951045)
Packaging Information
Specifications
Specification Test Results
- Assay:28 - 33 %
- Density (20/4):0.880 - 0.910
- Evaporation residue:Max. 100 ppm
- Heavy metals (as Pb):Max. 10 ppm
- Cl (Chloride):Max. 10 ppm
- CO3 (Carbonate):Max. 100 ppm
- SO4 (Sulphate):Max. 20 ppm
- Fe (Iron):Max. 1 ppm
- Pb (Lead):Max. 1 ppm
Specifications
- Pack type:Glass bottle
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



