Ethanol, GPR RECTAPUR®
Plastic drum, 25 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Caution: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.
Synonyms: Ethyl alcohol
- CAS Number:64-17-5
- Formula:C₂H₅OH
- MDL Number:MFCD00003568
- Molecular Weight:46.07 g/mol
- Boiling Pt:78.3 °C (1013 hPa)
- Melting Pt:–117 °C
- Density:0.7895 g/cm³ (20 °C)
- Flash Pt:12 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-578-6
- UN:1170
- ADR:3,II
- REACH:01-2119457610-43
Documents
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
20824_ETHANOL 96 % VOL GPR RECTAPUR_BSE_REV0
BSE/TSE Certificates
P20_VWRC20824.296_SPEC_EN.pdf P20_VWRC20824.321_SPEC_EN.pdf P20_VWRC20824.365_SPEC_EN.pdf P20_VWRC20824.467_SPEC_EN.pdf P20_VWRC20824.558_SPEC_EN.pdf
Product specification
More
VWR Chemicals Big Packs and Bulks VWR Denatured Ethanol
Product Literature
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.8 %
- Assay (V/V):96.0 - 96.3 %
- Acidity:Max. 0.0002 meq/g
- Aldehydes (as CH3CHO):Max. 15 ppm
- Evaporation residue:Max. 20 ppm
Specifications
- Pack type:Plastic drum
- Pk:25 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



