Order Entry
Italy
Orders LinkContactUsLinkComponent
Zinc
Plastic bottle for solids, 1 kg
  29074.294
Zinc
  29074.294
 :  7440-66-6
Zinc
Zinc
Plastic bottle for solids, 1 kg
 :  7440-66-6
1 * 1 kg
  •  :7440-66-6
  •  :Zn
  •  :MFCD00011291
  •  :65.39 g/mol
  •  :12,10255
  •  :908 °C (1013 hPa)
  •  :420 °C
  •  :7.14 g/cm³ (20 °C)
  •  :Ambient
  •  :231-175-3
  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg