Zinc
Plastic bottle for solids, 250 g
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
- CAS Number:7440-66-6
- Formula:Zn
- MDL Number:MFCD00011291
- Molecular Weight:65.39 g/mol
- Merck Index:12,10255
- Boiling Pt:908 °C (1013 hPa)
- Melting Pt:420 °C
- Density:7.14 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-175-3
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g



