Ethyl acetate ≥99%, TECHNICAL
Plastic bottle, 1 L
Supplier: VWR Chemicals
Ethyl acetate ≥99%, TECHNICAL
Catalog # 23879.295
Supplier: VWR Chemicals
CAS Number: 141-78-6
Jump to:
Product Details & Documents
Synonyms: Acetic acid ethyl ester
- CAS Number:141-78-6
- Formula:CH₃COOC₂H₅
- MDL Number:MFCD00009171
- Molecular Weight:88.11 g/mol
- Merck Index:13,03792
- Boiling Pt:77.1 °C (1013 hPa)
- Melting Pt:–83 °C
- Density:0.902 g/cm³ (20 °C)
- Flash Pt:–4 °C
- Storage Temperature:Ambient
- EINECS:205-500-4
- UN:1173
- ADR:3,II
- REACH:01-2119475103-46
Documents
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Packaging Information
P20_VWRC23879.295_SPEC_EN.pdf P20_VWRC23879.364_SPEC_EN.pdf P20_VWRC23879.466_SPEC_EN.pdf P20_VWRC23879.557_SPEC_EN.pdf spec_VWRC23879.295
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications
Specification Test Results
- Assay:Min. 99 %
- Appearance:Clear colourless liquid
- IR Spectrum:Passes test
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Cat. No.:23879.295
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



