Order Entry
Italy
Orders LinkContactUsLinkComponent
Accessories

Accessories

Gelatin filters, wavelength 546 nm, bandpass ± 5 nm
Catalog # 633-0117
Accessories
Catalog # 633-0117
Accessories

Gelatin filters, wavelength 546 nm, bandpass ± 5 nm

Price
638,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Specifications
  • Description:
    Gelatin filters, wavelength 546 nm, bandpass ± 5 nm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.