Order Entry
Italy
Orders LinkContactUsLinkComponent
VWR®, Clamps

VWR®, Clamps

8 - 35 mm
Catalog # 241-0037
Supplier:   VWR Collection
VWR®, Clamps
Catalog # 241-0037
Supplier:  VWR Collection
VWR®, Clamps
VWR®, Clamps

8 - 35 mm

Price
91,20 €
Log in for availability
1 * 3 pezzi
Product Details & Documents

Stainless steel. Easy to use, robust clamps.

  • Independently adjustable jaws, jaw height 16 and 25 mm
  • With two easy action knurled screws for adjusting each of the movable clamping jaws independently of each other

Documents

Specifications
  • Spanning width:
    8 - 35 mm
  • Seq:
    1
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.