Order Entry
Italy
Orders LinkContactUsLinkComponent
Vials with inserts, WHEATON®

Vials with inserts, WHEATON®

Autosampler vial, with 0,1 ml insert, 0,10 ml, 12×32 mm, Clear, No, Screw
  WHEA225195
Vials with inserts, WHEATON®
  WHEA225195
Vials with inserts, WHEATON®
Vials with inserts, WHEATON®

Autosampler vial, with 0,1 ml insert, 0,10 ml, 12×32 mm, Clear, No, Screw

 
493,00 €
   
1 * 100 pezzi
 

These plastic autosampler vials combine thermoplastic polymer outer vial shells with type I borosilicate glass inserts.

  • 100 µl glass insert
  • Available with snap/crimp or screw top finish
  • Conical interior ensures maximum retrieval of contents without the hassle of removable inserts

The thermoplastic polymer provides excellent dimensional tolerance reliability for capping and instrumentation use and safety from breakage.

 
  • Cap included:
    No
  • Ø×H:
    12×32 mm
  • Capacity:
    0,10 ml
  • Colour:
    Clear
  • Description:
    Autosampler vial, with 0,1 ml insert
  • Seq:
    02
  • Cap type:
    Screw