Order Entry
Italy
Orders LinkContactUsLinkComponent
Apparatus - Antonacopoulos
Apparatus - Antonacopoulos
Spare adapter, 70/50 NS, 29/32 NS
Catalog # LENZ05434002
Supplier:   Lenz Laborglas
Apparatus - Antonacopoulos
Catalog # LENZ05434002
Supplier:  Lenz Laborglas
Apparatus - Antonacopoulos
Spare adapter, 70/50 NS, 29/32 NS
Price
345,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Documents
Specifications
  • Cone size:
    70/50 NS
  • Socket size:
    29/32 NS
  • Description:
    Spare adapter
  • Seq:
    03