UV blocking eyewear
Spectacles (anti-UV)
Catalog # 730-1033
Supplier: UVP ULTRA VIOLET PRODUCTS
Jump to:
Product Details & Documents
Eye and face protection for applications involving short wave ultraviolet sources. Available in three styles.
- Spectacles are made of impact resistant PC; the top and side-shield design provides full UV protection
- Goggles are constructed of specially formulated plastic and feature a flexible rubber trim which fits comfortably on the face and over most prescription spectacles, allowing indirect ventilation
ANSI-Z87.1 1986
Certifications: ANSI-Z87.1 1986
Documents
UVPA98-0002-01_TDS_EN
Datasheet
Specifications
- Description:Spectacles (anti-UV)
- Seq:01